missing translation for 'onlineSavingsMsg'
Learn More

BD Protein A: HRP

Product Code. 15855459
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15855459 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15855459 Supplier BD Supplier No. 610438
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For routine western blotting analysis

  • Format: HRP
  • Storage buffer: 1mg/mL in 50% glycerol, 20mM sodium phosphate pH 7.5, 150 mM NaCl

Western blot

TRUSTED_SUSTAINABILITY

Specifications

Product Type HRP Protein A
Quantity 1 mg
Concentration 1 mg/mL
For Use With (Application) Western Blot
Content And Storage 4°C

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.