missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human GM-CSF

Product Code. 15896178 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896178 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15896178 Supplier BD Supplier No. 550068
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stimulates precursor cells of granulocytes, macrophages, and eosinophils

Quantitative standard for measuring human GM-CSF protein levels in a GM-CSF specific sandwich ELISA with the purified BVD2-23B6 antibody as capture antibody and the biotinylated BVD2-21C11 as detection antibody

  • Reactivity: Human
  • Storage buffer: Frozen aqueous buffered solution containing BSA
  • Biological Activity: Proliferation measured using TF-1 indicator cells
  • Specific Activity: 0.1- 1.0 x 109 Unit/mg
  • ED50: 10 - 90pg/ml

Bioassay, Enzyme-linked immunosorbent assay standard, Flow cytometry block

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 80°C
Quantity 10 μg
Source Human

Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.