missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human Interleukin-5

Product Code. 15817808 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15817808 5 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817808 Supplier BD Supplier No. 554606
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can be used as blocking control to demonstrate specificity of IL-5 staining by PE- or APC- labeled TRFK5 antibody or PE-labeled JES1-39D10 antibody

Interleukin-5 (IL-5) is the lymphokine responsible for activities attributed to eosinophil differentiating factor (EDF), B cell growth factor II (BCGFII) and T cell-replacing factor (TRF). IL-5 induces eosinophil differentiation and promotes eosinophil survival and activation. This is a 28 to 31 kD protein dimer, as determined by SDS-PAGE (in non-reducing conditions).

  • Supplied as frozen liquid with no preservatives
  • Purity: >95%
  • Reactivity: human
  • Concentration: 100μg/mL
  • Stored buffer: frozen aqueous buffered solution containing BSA
  • Endotoxin level: ≤0.1ngμg of human IL-5
  • Size: 5μg

Bioassay/ELISA standard/Intracellular block/flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 μg/mL
Endotoxin Level 0.1ng/μg
For Use With (Application) Blocking, ELISA, Flow Cytometry, Functional Assays, Immunochemistry, Immunochemistry
Purity >95%
Quantity 5 μg
Content And Storage -80°C
Reactivity Human
Format Frozen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.