missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human Interleukin-6 (IL-6)

Product Code. 15826188 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15826188 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15826188 Supplier BD Supplier No. 550071
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An absorbance assay based on the Beers-Lambert law

Quantitative standard for measuring human IL-6 protein levels in an IL-6 specific sandwich ELISA with purified MQ2-13A5 antibody as a capture antibody and biotinylated MQ2-39C3 as the detection antibody

  • Storage buffer: Frozen aqueous buffered solution containing BSA and glycerol
  • Biological Activity: Activity Range is measured in a proliferation assay using TF-1 indicator cells at 105cells/ml
  • Specific Activity: 0.5 - 3.0 x 108 Unit™/mg
  • ED50: 30- 200pg/ml; Observed dose response: 50 fold

Bioassay, Enzyme-linked immunosorbent assay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 10 μg
Source Human

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.