missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human Interleukin-7 (IL-7)

Product Code. 15827808 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μ g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15827808 5 μ g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15827808 Supplier BD Supplier No. 554608
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent lymphoid cell growth factor which affects pre-B, pro-B, and early T cells

Lyophilized from a solution comprised of 0.22 ++m sterile-filtered PBS and containing 50μg bovine serum albumin per ++g of cytokine

  • Reactivity: Human
  • Storage Buffer: Lyophilized in an aqueous buffered solution containing BSA

Enzyme-linked immunosorbent assay standard, bioassay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 5 μ g
Source Human

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.