missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Mouse GM-CSF

Product Code. 15897788
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15897788 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15897788 Supplier BD Supplier No. 554586
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Has potent stimulatory effects on growth and differentiation of bone marrow progenitor cells

  • Produced by activated T cells, macrophages, vascular endothelial cells and fibroblasts
  • Contains 124 amino acid residues
  • Helps in generating granulocytes, monocytes/macrophages, and megakaryocytes
  • Act on mature leukocytes to promote inflammatory responses in peripheral tissues
  • Appears as 15.5 - 19kD band in reducing and non-reducing SDS-PAGE analysis
  • Storage buffer: frozen aqueous buffered solution containing BSA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 10 μg
Source Mouse

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.