Learn More
Gibco™ Resurge™ CD Pak
Description
Resurge CD Pak are a diverse set of chemically defined supplement formulations to complement a variety of base media. The CD PAK contains one of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each. The supplements have been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations. Therefore, these two components should be added by users and monitored as appropriate.
Specifications
Specifications
| Content And Storage | •One of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each. •Storage conditions: 2° to 8°C. •Store in cool, dry conditions in well-sealed containers. |
| Cell Line | CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free |
| Form | Powder |
| Product Type | Media Supplement |
| Format | Bottles |
| Product Line | Resurge |
| Quantity | 1 Ea. |
| Source | Animal origin-free |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.