missing translation for 'onlineSavingsMsg'
Learn More

BD Stemflow™ Human Neural Lineage Analysis Kit

Product Code. 15801109 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15801109 25 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15801109 Supplier BD Supplier No. 561526
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Sox2 PerCP-Cy™5.5, GFAP Alexa Fluor™ 647, Doublecortin PE, Nestin Alexa Fluor 647, Ki-67 Alexa Fluor 488, Sox1 PerCP-Cy5.5, CD44 FITC; BD Cytofix™ fixation buffer; BD Phosflow™ perm buffer III

Provides a comprehensive research system for the reliable, in-depth characterization of differentiation stages of human neural stem cells (hNSCs) from a heterogeneous culture

  • Provides intracellular, multimarker data with antibodies that can be used at various time points to resolve hNSCs from neurons and glia in a heterogeneous population
  • Includes key neural differentiation markers CD44, doublecortin, nestin, Sox1, Sox2, and GFAP, plus Ki-67 for identification of cycling cells
  • Delivers a streamlined solution for consistent experiments with fluorochrome conjugated antibodies, buffers and a detailed protocol
  • Regulatory Status: RUO

TRUSTED_SUSTAINABILITY

Specifications

Reactivity Human
Quantity 25 Tests
For Use With (Application) Flow Cytometry
Content And Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.