missing translation for 'onlineSavingsMsg'
Learn More

BD PAXgene™ Blood RNA Tube

Product Code. 12937706
Change view
Click to view available options
Capacity (Metric):
2.5 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12937706 2.5 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12937706 Supplier BD Supplier No. 762165
  • ONLINE EXCLUSIVE PRICE
    8403.56 €
    3456.00 € / Pack of 100
    Save 4947.56 €
    58% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers a convenient and closed system for the collection, storage and transportation of whole blood that requires intracellular RNA stabilization. BD Vacutainer™ PAXgene™ Blood RNA Tube is for in vitro diagnostic use and is CE-marked.

The collection of whole blood is the first step in many molecular assays used to study intracellular RNA. A major challenge in this type of testing is the instability of intracellular RNA, which rapidly degrades within hours after blood collection. Furthermore, certain species of RNA, through the process of gene induction, increase in vitro after blood collection. Both in vitro RNA degradation and gene induction can lead to an under- or over-estimation of in vivo relative gene transcript number.

The PAXgene™ Blood RNA Tube contains an additive that stabilizes the in vivo gene transcription profile by reducing in vitro RNA degradation and minimizing gene induction. When used in conjunction with the PAXgene™ Blood RNA Kit, the Blood RNA Tube provides for accurate detection and quantitation of gene transcripts. Store unused PAXgene Blood RNA Tubes at 18–25°C. (Limited excursion temperatures up to 40°C are permitted.)

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Product Type Cell Preparation Tube
Additive PAXgene Blood RNA Preservative Solution, 6.9mL
Material PET
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 2.5 mL
Includes 100/case
For Use With (Application) Molecular Diagnostic Testing
Label Type Paper
Length (Metric) 100 mm
Diameter (Metric) Outer 16 mm
Certifications/Compliance CE marked
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • PAXgene Blood RNA Tube
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Tetradecyltrimethylammonium oxalate, (+)-Tartaric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.