Learn More
BOLA Beakerliner
Product Code. p-200109957
Click to view available options
Height (Metric):
8.5 mm
21 mm
29 mm
32 mm
48 mm
50 mm
Unit Size:
Each
Product Code. 15319986
Brand: BOLA C36202
Only null left
Add to basket
Add to basket
Beakerliner Stirring magnetic core made of Alnico 5.
PTFE-encapsulated magnetic core (Alnico 5) axially mounted in a guide cage made of PTFE, universal chemical resistance.
Specifications
Autoclavable Autoclavable
Color White
Height (Metric) 8.5 mm
Coating PTFE