missing translation for 'onlineSavingsMsg'
Learn More

BEBT-908, MedChemExpress

Product Code. 16477706
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477706 100 mg 100mg
16470358 5 mg 5mg
16437898 10 mg 10mg
16403847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477706 Supplier Medchem Express Supplier No. HY19763
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BEBT-908 (PI3Kα inhibitor 1) is a selective PI3Kα inhibitor extracted from patent US/20120088764A1, Compound 243, has an IC5050≤1 μM) .

BEBT-908 (PI3Kα inhibitor 1) is a selective PI3Kα inhibitor extracted from patent US/20120088764A1, Compound 243, has an IC5050≤1 μM) .

Specifications

Chemical Name or Material BEBT-908
CAS 1235449-52-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H25N9O3S
Quantity 100 mg
Synonym PI3Kα inhibitor 1
Solubility Information DMSO : ≥ 37 mg/mL (72.90 mM)
SMILES CN(CC1=CC(N=C(C2=CN=C(NC)C=C2)N=C3N4CCOCC4)=C3S1)C5=NC=C(C(NO)=O)C=N5
Molecular Weight (g/mol) 507.57
Formula Weight 507.57
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.