missing translation for 'onlineSavingsMsg'
Learn More

Beclabuvir, MedChemExpress

Product Code. p-200100256
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495158

Brand: Medchem Express HY12429

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Beclabuvir is an allosteric inhibitor that binds to thumb site 1 of the hepatitis C virus (HCV) NS5B RNA-dependent RNA polymerase, and inhibits recombinant NS5B proteins from HCV genotypes 1, 3, 4, and 5 with IC50 of < 28 nM.

Beclabuvir is an allosteric inhibitor that binds to thumb site 1 of the hepatitis C virus (HCV) NS5B RNA-dependent RNA polymerase, and inhibits recombinant NS5B proteins from HCV genotypes 1, 3, 4, and 5 with IC50 of < 28 nM.

Specifications

Chemical Name or Material Beclabuvir
CAS 958002-33-0
Color White
Purity Grade Notes Research
Molecular Formula C36H45N5O5S
Quantity 10 mg
Synonym BMS-791325
Solubility Information DMSO : ≥ 30 mg/mL (45.47 mM)
SMILES O=C(N1C2CCC1CN(C)C2)[C@@]3(CN4C5=C(C6CCCCC6)C7=CC=C(C(NS(N(C)C)(=O)=O)=O)C=C47)[C@H](C8=CC(OC)=CC=C85)C3
Molecular Weight (g/mol) 659.84
Formula Weight 659.84
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.