missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclomethasone Dipropionate, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Beclometasone Dipropionate |
| CAS | 5534-09-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 H37 Cl O7 |
| Quantity | 250 mg |
| Synonym | 9-Chloro-11beta-hydroxy-16beta-methyl-3,20-dioxopregna-1,4-diene-17,21-diyl Dipropanoate, Beclometasone Dipropionate, Betamethasone Dipropionate Imp. E (Pharmeuropa), (11β,16β)-9-Chloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate, 9α-Chloro-16β-methylprednisolone 17,21-dipropionate, Aerobec, Aldecin, Aldecin AQ nasal, Anceron, Andion, Beclacin, Beclate, Beclazone, Beclazone 250, Beclazone 50, Beclomet, Beclometasone 17,21-dipropionate, Beclometasone dipropionate, Beclomethasone 17,21-dipropionate, Beclomethasone 17α,21-dipropionate, Beclomethasone dipropionate, Beclotide, Beclotide 100, Becloval, Beclovent, Beclovent Inhaler, Becodisks, Beconase, Beconase AQ, Becotide, Belchlorhinol, Belcoforte, Belcomet, Clenil A, Entyderma, Inalone O, Inalone R, Korbutone, Propaderm, Propaderm Forte, QVAR, Qvar 50, Rino-Clenil, Sanasthmax, Sanasthmyl, Sanasthymyl, Sch 8020W, Vancenase, Vancenase AQ, Vanceril, Vanceril DS, Ventolair, Viarex, Viarox, orBec |
| InChI Formula | InChI=1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1 |
| SMILES | CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate |
| Molecular Weight (g/mol) | 521.04 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction