missing translation for 'onlineSavingsMsg'
Learn More

Behenic Acid, TRC

Product Code. 30332686
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332686 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332686 Supplier Toronto Research Chemicals Supplier No. TRCB11958010G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Behenic Acid
CAS 112-85-6
Molecular Formula C22 H44 O2
Quantity 10 g
Synonym Docosanoic Acid, 1-Docosanoic acid, B 95, B 95 (acid), Behenic acid, EXL 5, Edenor C 22-85R, Edenor C 22-85RGS, Glycon B 70, Hydrofol 2022-55, Hydrofol Acid 560, NAA 222S, NAA 22S, NSC 32364, Prifac 2987, Prifrac 2989, n-Docosanoic acid
InChI Formula InChI=1S/C22H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H,23,24)
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)O
IUPAC Name docosanoic acid
Molecular Weight (g/mol) 340.58
Formula Weight 340.3341
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.