missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil

Product Code. 11737927 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (English):
13 to 20 gal.
2 to 4 gal.
20 to 30 gal.
40 to 55 gal.
5 to 9 gal.
Unit Size:
Pack of 100
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) unitSize
11737927 20 to 30 gal. Pack of 200
11392154 2 to 4 gal. Pack of 200
11302164 5 to 9 gal. Pack of 200
11312164 13 to 20 gal. Pack of 200
10296674 40 to 55 gal. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11737927 Supplier Bel-Art™ Supplier No. F131643138
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.

  • Bright red polypropylene bags include black biohazard symbol
  • Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
  • Sterilization patch darkens when subjected to steam sterilization
  • High temperature steam autoclavable to 135°C (275°F)

Specifications

Description 31W x 38H in. (79 x 97cm)
Autoclavable Autoclavable
Certifications/Compliance ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact
Closure Type Zip Tie
Color Red
Disposable Single-use
For Use With (Application) Used for biohazard waste
Includes Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish
Material Polypropylene
Sterility Non-sterile
Thickness (English) 1.50 mil
Dimensions (L x W) 31 x 38 in.
Length (English) 38 in.
Width (English) 31 in.
Height (English) 38 in.
Height (Metric) 97 cm
Product Type Biohazard Disposal Bag
Capacity (English) 20 to 30 gal.
Capacity (Metric) 95 L
Length (Metric) 97 cm
Width (Metric) 79 cm
Thickness (Metric) 0.279 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.