missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Conical Tube Holders, 50 mL

Product Code. 11332164 Shop All SP Scienceware Products
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11332164 Blue Pack of 5
10296354 White Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332164 Supplier Bel Art™ Supplier No. F187950001
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Leave both hands free for manipulation of 50 mL sample tubes

Leave both hands free for manipulation of 50 mL sample tubes

  • Handy polysterene base securely holds a 50 mL conical centrifuge tube in place, freeing hands for tasks such as pipetting and adjustment of instrumentation
  • Allow better control of procedures involving measurement, analysis and modification
  • Grip Style securely holds the tube in the base and moves with the tube when it is lifted from the benchtop; remove tubes by holding the base and pulling out tube
  • Non-Grip Style allows the tube to be lifted freely from the base

Specifications

Autoclavable Not Autoclavable
Color Blue
Holds Tubes (5 mL)
Material Polystyrene
Positions 1
Shape Conical
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) Conical tube
Product Type Test Tube Single Rack
Shelves None
Array 1 x 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.