missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Sharp Object Safety Pouch

Product Code. 11782225
Change view
Click to view available options
Capacity (Metric):
1100 cc
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11782225 1100 cc Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11782225 Supplier Bel-Art™ Supplier No. H132340000
  • ONLINE EXCLUSIVE PRICE
    232.00 €
    229.51 € / Pack of 200
    Save 2.49 €
    1% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers protection against cuts by contaminated sharp objects during work in labs, clinics, and medical offices

  • Pouch remains open in an oval shape by folding the bottom flaps
  • After filling, the top flaps close and lock in place for safe disposal
  • Solid paperboard pouch is 10 mil thick resists puncture by sharps
  • Printed with bold biohazard symbol
  • Measures 14 x 33 cm (5.5 x 13 in.) flat

Order Info

Safety pouch stand (22-197-061 ) sold separately.

Specifications

Capacity (Metric) 1100 cc
Capacity (English) 0.29 gal.
Color White
Autoclavable Not Autoclavable
For Use With (Application) For collection and disposal of sharp objects
Material Corrugated Cardboard
Product Type Sharps Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.