Learn More
Bel-Art™ SP Scienceware™ Blow-Hard™ O.S. Extra™ Dust Removing Spray
Fast and easy cleaning for hard-to-reach areas
Brand: Bel-Art™ F17080-0200
Code : NEW
Additional Details : CAS Number : 811-97-2 Weight : 0.36900kg Transport : UN number : 1950 Chem class : 2.2
Description
- Provides powerful blasts of triple-filtered air for cleaning laboratory equipment too fragile for wiping
- Included extension tube penetrates difficult corners and tight spaces
- Ozone-safe and nonflammable
- Warning instructions for safe use printed on can
Specifications
Dust Removing Spray | |
No | |
10 oz. |
25.3 x 14.1 x 14.2 in. (64.2 x 35.8 x 36.1 cm) | |
For cleaning a wide variety of laboratory equipment which could be damaged by wiping | |
0.29 L |
Safety and Handling
- Blow-Hard O.S. Extra
Signal Word
- Warning
Hazard Category
- Gases under pressure
Hazard Statement
- H280-Contains gas under pressure; may explode if heated.
- EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
- P410+P403-Protect from sunlight Store in a well-ventilated place.
Supplemental information
- MIXTURE LIST-Contains : norflurane