missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves

Product Code. p-144199 Shop All SP Scienceware Products
Click to view available options
Length (English) Cuff:
11 in.
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 10762682

Brand: Bel-Art™ H132010001

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).

  • Ideal for removing hot objects from an autoclave or oven
  • Heat-resistant up to 232°C
  • Replaces hazardous asbestos gloves
  • Extended gauntlet cuff available for additional forearm protection
  • Gloves are not waterproof and should not be used to handle dry ice
  • Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer

Specifications

Certifications/Compliance CE
Accelerator Free Yes
Applications Laboratory, Industrial
Coating Material Uncoated
Color Orange
Latex Free Yes
Material Terrycloth
Product Type Autoclave Gloves
Size Universal
Style Full-hand Mitt
Temperature (English) Max. 450°F
Temperature (Metric) Max 232°C
For Use With (Application) Handling Hot Objects
Glove Cuff Style Gauntlet
Series H13201
Length (English) 18.5 in.
Length (English) Cuff 11 in.
Length (Metric) 47 cm
Length (Metric) Cuff 28 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.