missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Conical Tube Holder

Product Code. 11765234 Shop All SP Scienceware Products
Click to view available options
Positions:
1
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 11765234

Brand: Bel-Art™ F187960000

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Allows user to clearly view the entire contents of tubes. Bel-Art™ Conical Tube Holder sets up in just seconds to hold either a 15 ml or a 50 ml conical tube.

Allows user to clearly view the entire contents of tubes. Bel-Art™ Conical Tube Holder sets up in just seconds to hold either a 15 ml or a 50 ml conical tube.

  • Versatile holder supports either a 15 or 50mL conical tube while allowing a view of the entire tube contents
  • Move tube and rack with one hand by pinching the uprights
  • Insert or remove tube from the holder with one hand
  • Steam autoclavable at 121°C (250°F)
  • Chemical-resistant polypropylene

Specifications

Product Type Test Tube Single Rack
Array 1 x 1
Autoclavable Autoclavable
Color Transparent
Holds Tubes (15 or 50 mL)
Material Polypropylene
Positions 1
Shape Circular
Shelves None
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) 15 and 50mL Conical Tubes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.