missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Labeled Wash Bottles

Product Code. 30193464 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
500 mL
For Use With (Application):
Acetone
Isopropanol
Water
Conditionnement:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Capacity (Metric) For Use With (Application) Conditionnement
30193464 500 mL Isopropanol Pack of 6
11592463 500 mL Acetone Pack of 6
11572463 500 mL Water Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
All bottles are intended for dispensing use only, not for long-term storage. Use LYOB only with solvents compatible with LDPE.
Code produit 30193464 Fournisseur Fisherbrand™ Code fournisseur F116460624
  • ONLINE EXCLUSIVE PRICE
    49.70 €
    40.23 € / Pack of 6
    Save 9.47 €
    19% Off
Afficher toutes les promotions
 Restricted Item
Estimated Shipment: 05-01-2027
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
All bottles are intended for dispensing use only, not for long-term storage. Use LYOB only with solvents compatible with LDPE.

Includes

Safety labeled wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemmical Abstract Service (CAS), protective equipment symbols and target organ information.

Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.

  • Easy-to-read printed bottles
  • Wide mouth 53 mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
  • Toluene is red LDPE bottle
  • Polypropylene closure with leakproof design
  • Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol

Compliance

OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Spécification

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Color Natural
For Use With (Application) Isopropanol
Material LDPE, Polyethylene
Sterility Non-sterile
Closure Color Yellow
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Chemical Label Isopropanol
Closure Material Polypropylene
Closure Size 53 mm
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Cylindrical
Disposable Reusable
Product Type Safety-labeled Wash Bottle
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.