missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Mini Tubing Clamp

Use these small clamps for positive shutoff with fingertip control. Bel Art™ SP Scienceware™ Mini Tubing Clamp is durable, will not rust or corrode.

Brand:  Bel-Art™ F18227-0000

Code : A0

Additional Details : Weight : 0.10080kg

Product Code. 11744615

  • 37.30 € / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • Single-position on/off
  • No distortion, rust or corrosion
  • Use for thin-walled tubing under 4.7mm (0.19 in.) O.D.

Specifications

Specifications

Autoclavable
Mini Tubing Clamp
Acetal Plastic
Thin Wall Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers