missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Pipette Pump™ Pipetters

Product Code. 11727924 Shop All SP Scienceware Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
Volume (Metric):
0.2 mL
2 mL
10 mL
25 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) unitSize
11727924 Green 10 mL Each
11717924 Blue 2 mL Each
11707924 Yellow 0.2 mL Each
11727854 Red 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11727924 Supplier Bel-Art™ Supplier No. F378980000
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Provides safe, accurate and trouble-free pipetting. Bel Art™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.

  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Green
Disposable Reusable
For Use With Pipette
Grip Type Ergonomic
Material Polypropylene
Model Pipette Pump Pipettors
Number of Channels 1
Sterility Non-sterile
Type Pipette
Volume (Metric) 10 mL
For Use With (Equipment) Pipets
Resistance Acid and Alkali
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.