missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ SP Scienceware™ PLUM™ Emergency Covered Eye Wash Station

Product Code. 12739706
Change view
Click to view available options
Includes:
One 500 mL bottle of salen and one 200 mL bottle pH neutralizing solution, instructional pictogram, mirror and form fitting eye cups
Capacity (Metric):
500 mL, 200mL
Rinse Time:
5 min./500 mL Bottle, 2 min./200 mL Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Capacity (Metric) Rinse Time unitSize
12739706 One 500 mL bottle of salen and one 200 mL bottle pH neutralizing solution, instructional pictogram, mirror and form fitting eye cups 500 mL, 200mL 5 min./500 mL Bottle, 2 min./200 mL Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12739706 Supplier Bel Art™ Supplier No. F248805003

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SP Scienceware™ PLUM™ Emergency Covered Eye Wash Station offers instant access to fast, safe, and effective first aid for eye and skin injuries.

Suitable for high dust/dirt areas

  • Convenient, wall-mountable stations
  • Bottles are sterile filled and sealed and require no maintenance or cleaning
  • Stations feature clear instructional pictograms
  • Flexible, form-fitting eyecups ensure gentle and efficient rinsing
  • Solitions have three year shelf life from date of manufacture

Compliance

Meet all U.S. federal regulations. Comply with ANSI™ standards. NDC# and expiration date printed on every bottle.

Specifications

Capacity (English) 16 oz., 6.76 oz.
Capacity (Metric) 500 mL, 200mL
Certifications/Compliance ANSI Z358.1-2009, U.S. federal regulations
For Use With (Application) For Workplaces where there is a risk of Eye Injuries from Acids or Alkalis
Includes One 500 mL bottle of salen and one 200 mL bottle pH neutralizing solution, instructional pictogram, mirror and form fitting eye cups
Mounting Wall
Product Type Covered Double Eyewash Station
Rinse Time 5 min./500 mL Bottle, 2 min./200 mL Bottle
Sterility Sterile
Storage Requirements 5°C to 35°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.