missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ PLUM™ Emergency Eyewash Station Refills

Product Code. 12719706
Change view
Click to view available options
Product Type:
Saline Eyewash Solution Refill
Includes:
Three 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions
Capacity (Metric):
500 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Includes Capacity (Metric) Unit Size
12719706 Saline Eyewash Solution Refill Three 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions 500 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12719706 Supplier Bel Art™ Supplier No. F248800053

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Refill kits for PLUM station

Refill kits for PLUM station

  • Bottles contain a sterile 0.9% sodium chloride solution (saline) for rinsing dust and debris, or a 4.9% sterile phosphate buffer (pH neutralizing) solution to quickly neutralize acids and alkalis
  • Bottles have clear first aid instructions, twist-open design for quick access and a built-in eye cup with dust cap
  • Use to refill eye wash stations or take to job sites as part of first aid kits
  • Bottles require no maintenance and have a three-year shelf life from date of manufacture

Compliance

Meet all U.S. federal regulations. Comply with ANSI™ standards. NDC# and expiration date printed on every bottle.

Specifications

Capacity (English) 16 oz.
Capacity (Metric) 500 mL
For Use With (Equipment) PLUM Eyewash Stations or Alone
Includes Three 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions
Product Type Saline Eyewash Solution Refill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.