missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Polypropylene Sterilizing Trays

Product Code. 11745074
Change view
Click to view available options
Capacity (Metric):
5 L
13 L
18 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11745074 5 L Each
11762555 18 L Each
11752555 13 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11745074 Supplier Bel Art™ Supplier No. H162600000
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Trays are steam autoclavable at 121°C (250°F)
  • Covers are sold separately

Order Info

Covers are sold separately.

Specifications

Capacity (Metric) 5 L
Dimensions (L x W x H) 12 x 8 x 5 in. (30.5 x 20.3 x 12.7 cm)
Color White
Height (Metric) 12.7 cm
Length (Metric) 30.5 cm
Material Polypropylene
Type Sterilizing Tray
Width (Metric) 20.3 cm
For Use With Fits 5L Tray Cover, Mfr. No. H162590000
Length (English) 12 in.
Height (English) 5 in.
Shape Rectangular
Width (English) 8 in.
Capacity (English) 1.3 gal.
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.