missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Polypropylene Sterilizing Trays

Product Code. p-65234 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
5 L
13 L
18 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11745074

Brand: Bel Art™ H162600000

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Trays are steam autoclavable at 121°C (250°F)
  • Covers are sold separately

Order Info

Covers are sold separately.

Specifications

Capacity (Metric) 5 L
Dimensions (L x W x H) 12 x 8 x 5 in. (30.5 x 20.3 x 12.7 cm)
Color White
Height (Metric) 12.7 cm
Length (Metric) 30.5 cm
Material Polypropylene
Type Sterilizing Tray
Width (Metric) 20.3 cm
For Use With Fits 5L Tray Cover, Mfr. No. H162590000
Length (English) 12 in.
Height (English) 5 in.
Shape Rectangular
Width (English) 8 in.
Capacity (English) 1.3 gal.
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.