Learn More
Bel-Art™ Safety Cutter
Product Code. 16420745 Shop All SP Scienceware Products
Click to view available options
Blade Material:
Ceramic
Unit Size:
Pack of 3
Product Code. 16420745
Brand: Bel-Art™ F210120000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Easily and safely cut LabMat™ liners, plastic clamshell packaging, shrink wrap, paper, and more.
  • Faster and safer than scissors or utility knives: will not cut fingers
  • Ceramic blade stays rust-free and remains sharp longer than stainless steel blades
  • Blade does not need to be covered for safety
  • Elongated tear-drop shape measures 2.37 x 1.18 x 0.18 in. (6 x 3 x 0.5cm)
Specifications
Blade Material Ceramic
Product Type Safety Cutter
For Use With (Application) Easily cuts LabMat™ liner, clamshell packaging, Plastic bags, Shrink-wrap, Paper and much more
Size 2.38 x 1.18 x 0.18 in.
Sterility Non-sterile