missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Secador™ Auto Desiccator Cabinet, Vertical Model

Product Code. 11846890
Change view
Click to view available options
Color:
Amber
Clear
Cabinet Style:
Vertical
Vertical (Tinted)
Voltage:
230 V
Volume (English):
1.16 cu. ft.
1.55 cu. ft.
1.87 cu. ft.
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Cabinet Style Voltage Volume (English) Unit Size
11846890 Amber Vertical (Tinted) 230 V 1.87 cu. ft. Each
12319005 Clear Vertical 230 V 1.55 cu. ft. Each
15177823 Amber Vertical (Tinted) 230 V 1.16 cu. ft. Each
12309005 Clear Vertical 230 V 1.16 cu. ft. Each
11802742 Clear Vertical 230 V 1.87 cu. ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11846890 Supplier Bel-Art™ Supplier No. F420741228
  • ONLINE EXCLUSIVE PRICE
    1630.00 €
    1277.36 € / Pack of 1
    Save 352.64 €
    21% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A UV light blocking, dry environment with automatic electronic dehumidification.

A UV light blocking, dry environment with automatic electronic dehumidification.

  • Attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants.
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds.
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals.
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf.
  • 4.0 horizontal has a wider footprint, lower height and pull down oven style door.
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626.
  • 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info).
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in. H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H

Order Info

Replacement shelves are available.

Specifications

Autoclavable Not Autoclavable
Color Amber
Cabinet Style Vertical (Tinted)
Material Copolyester
Product Type Desiccator
Door Style Side-swinging
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 4.0
No. of Shelves 3
Resistance Chemicals, stains and fine cracks
Shelf Type Vented Adjustable
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5°C to 40°C
Volume (English) 1.87 cu. ft.
Dimensions (L x W) 16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English) 16.3 in.
Length (Metric) 41.4 cm
Width (English) 13.4 in.
Width (Metric) 34.1 cm
Height (English) 20.4 in.
Height (Metric) 51.8 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Voltage 230 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.