missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Secador™ Auto Desiccator Cabinet, Vertical Model

Artikelnummer. 12319005
Ansicht ändern
Click to view available options
Color:
Amber
Clear
Cabinet Style:
Vertical
Vertical (Tinted)
Voltage:
230 V
Volume (English):
1.16 cu. ft.
1.55 cu. ft.
1.87 cu. ft.
Packungsgröße:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Color Cabinet Style Voltage Volume (English) Packungsgröße
12319005 Clear Vertical 230 V 1.55 cu. ft. Each
15177823 Amber Vertical (Tinted) 230 V 1.16 cu. ft. Each
11846890 Amber Vertical (Tinted) 230 V 1.87 cu. ft. Each
12309005 Clear Vertical 230 V 1.16 cu. ft. Each
11802742 Clear Vertical 230 V 1.87 cu. ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 12319005 Lieferant Bel Art™ Lieferanten-Nr. F420731220
  • ONLINE EXCLUSIVE PRICE
    1240.00 €
    1167.41 € / Pack of 1
    Save 72.59 €
    5% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 29-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A UV light blocking, dry environment with automatic electronic dehumidification.

A UV light blocking, dry environment with automatic electronic dehumidification.

  • Attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants.
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds.
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals.
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf.
  • 4.0 horizontal has a wider footprint, lower height and pull down oven style door.
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626.
  • 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info).
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in. H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H

Bestellinformation

Replacement shelves are available.

Spezifikation

Autoclavable Not Autoclavable
Color Clear
Cabinet Style Vertical
Material Copolyester
Product Type Desiccator
Door Style Side-swinging
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 3.0
No. of Shelves 3
Resistance Chemicals, stains and fine cracks
Shelf Type Removable Perforated
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5°C to 40°C
Volume (English) 1.55 cu. ft.
Dimensions (L x W) 16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English) 16.3 in.
Length (Metric) 41.4 cm
Width (English) 13.4 in.
Width (English) Door 8.9 in.
Width (Metric) 34.1 cm
Width (Metric) Door 22.5 cm
Height (English) 16.4 in.
Height (English) Opening 12.3 in.
Height (Metric) 41.7 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Voltage 230 V
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.