missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Secador™ Gas-Purge Desiccator Cabinet

Product Code. 31337577 Shop All SP Scienceware Products
Click to view available options
Height (Metric):
21.3 cm
41.7 cm
51.8 cm
Color:
Amber
Clear
No. of Shelves:
1 Fixed Vented Adjustable
2 vented adjustable shelves
3
3 perforated removable shelves
Model:
1.0
3.0
4.0
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 31337577

Brand: Bel Art™ F420740002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Available in a wide range of sizes for gas-purged desiccation.

Available in a wide range of sizes for gas-purged desiccation.

  • Create a controlled environment with dry nitrogen desiccation or other gases of choice; Dust-free and desirable for many applications including clean room environments
  • Rear mounted gas port valves accept 0.25 in. tubing and can be closed to use standard desiccants
  • The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products; Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing, and chemical attack
  • Amber color models reduce visible light penetration by over 50% for optimal storage light-sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • Models 1.0, 2.0, 3.0 and 4.0: Vertical units have a compact 16.3 L x 13.4 in.W footprint for space-saving stacking up to 3 units high and side swinging door that opens to the left; 4.0 horizontal has a wider footprint, lower height and pull down oven style door
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626

Dimensions

  • Secador 1.0: 0.74 cu. ft. internal volume, 16.3 L x 13.4 W x 8.4 in. H
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in.H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H

Order Info

Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100. Replacement shelves available: 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves; Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001.

Specifications

Autoclavable Not Autoclavable
Color Clear
Cabinet Style Horizontal
Material Copolyester
Product Type Desiccator
Door Style Bottom-hinged and gasketed
For Use With (Application) Create a controlled environment with dry nitrogen desiccation or other gases of choice, dust-free and desirable for many applications including clean room environments
Holds 70 Cans
Includes Rear mounted gas port valves, carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 4.0
No. of Shelves 2 vented adjustable shelves
Resistance Chemicals, stains and fine cracks
Volume (English) 1.87 cu. ft.
Width (English) 13.4 in.
Width (English) Door 8.9 in.
Width (Metric) 34.1 cm
Width (Metric) Door 22.5 cm
Height (English) 20.4 in.
Height (English) Opening 16.4 in.
Height (Metric) 51.8 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.