missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Secador™ Standard Desiccator Cabinet, Vertical Model

Product Code. 10551712 Shop All SP Scienceware Products
Click to view available options
Height (Metric):
31.4 cm
41.7 cm
51.8 cm
Color:
Amber
Blue with Clear Door
Clear
Clear with Blue Ends
No. of Shelves:
2
3
Volume (English):
1.17 cu. ft.
1.58 cu. ft.
1.9 cu. ft.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10551712

Brand: Bel Art™ F420741006

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Constructed of Durastar™ co-polyester. Bel-Art™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing, and chemical attack.

Constructed of Durastar™ co-polyester. Bel-Art™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing, and chemical attack.

  • Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Compact 34.1 W x 41.4cm D footprint for space saving stacking up to 3 units high
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
  • Can be used with desiccant cartridges, F42049-0100 or F42046-0000

Bestellinformation

Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. F42049-0100, F42046-0000

Spezifikation

Autoclavable Not Autoclavable
Color Clear with Blue Ends
Cabinet Style Vertical
Material Copolyester
Product Type Desiccator
Door Style Left-hinged and gasketed
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 4.0
No. of Shelves 3
Resistance Chemicals, stains and fine cracks
Shelf Type Removable Perforated
Volume (English) 1.9 cu. ft.
Width (English) 13.4 in.
Width (English) Door 8.9 in.
Width (Metric) 34.1 cm
Width (Metric) Door 22.5 cm
Height (English) 20.4 in.
Height (English) Opening 16.4 in.
Height (Metric) 51.8 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt