missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Spinbar™ Magnetic Sink Strainer

Product Code. 11823210 Shop All SP Scienceware Products
Change view
Click to view available options
Color:
Blue
Green
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11823210 Blue Each
11833210 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11823210 Supplier Bel-Art™ Supplier No. F377870000
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Resistant to chemicals sink strainer has four strong magnets in each corner to attract and hold Spinbar magnetic stirring bars.

  • Long slits in the sides and skirt allow fluids to drain easily and rapidly
  • A wide skirt enables strainer to be used in a variety of sink drains; flat edge on one side allows it to be placed close to the sink wall
  • Four legs on the underside anchor the strainer firmly in sink drain and prevent it from being dislodged by flowing liquid
  • Color choices improve visibility of stirring bars

Specifications

Color Blue
Dimensions (L x W x H) 5 x 1.44 in.
For Use With (Application) Prevents stir bar from washing down drain
For Use With (Equipment) Stirring bars
Includes 4 strong magnets in each corner, long slits in the sides and wide skirts
Diameter (Metric) 127 mm
Length (Metric) 127 mm
Material PTFE, Alnico Magnet
Quantity 1
Shape Disc
Type Magnetic Stir Bar Arrester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.