Learn More
Bel-Art™ Touch Free™ Automatic Waste Cans
Automatically open and close without a touch, protecting users from waste receptacle contact
160.14 € - 229.00 €
Specifications
Includes | Brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications) |
---|
Includes: Each unit is supplied with a brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications).
Product Code | Brand | Capacity (English) | Color | Dimensions (L x W x H) | Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Color | Dimensions (L x W x H) | Material | Price | Quantity & Availability | ||
12300629
|
Bel-Art™
F13202-0022 |
7.3 gal. | Red | 36.8 x 26.7 x 57.1 cm (14.4 x 10.5 x 22.5 in.) | ABS |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11728414
|
Bel-Art™
F13202-0020 |
8.7 gal. | Gray | 37.2 x 26.5 x 65.4 cm (14-5/8 x 10-1/2 x 25-3/4 in.) | Plastic |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11738414
|
Bel-Art™
F13202-0030 |
12 gal. | Silver | 36.8 x 27 x 67.3 cm (14.5 x 10.5 x 26.5 in.) | Stainless Steel |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Automatically open and close without a touch, protecting users from waste receptacle contact
- When a hand or solid object is moved over the top, waste can detects motion, opens lid to accept waste, and then closes automatically
- Fitted well helps to reduce odor and airborne contamination
- Durable, dependable construction accommodates Scienceware™ liners, biohazard liners or commercially available trash bags
- ON-OFF switch deactivates AUTO-OPEN/CLOSE function
- Biohazard bags sold separately
2 Gallon (Cat. No. 11-394-451)
- Ideal for benchtop use
- Removable rigid autoclavable polypropylene liner will not leak or rust
- Body and lid made from easy-to-clean, molded ABS plastic
- White pail with white top
- Line with 14 x 19 in. biohazard disposal bags or suitable size standard trash bags
- Pail made from polypropylene with an ABS plastic top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- Gray pail with gray top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- Pail made from polypropylene with an ABS plastic top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- Red pail with red top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- Small footprint enables a fit into tight spaces
- Stainless-steel pail with black ABS plastic top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
Prior to use, cans should be appropriately labeled with one of the included labels to indicate the type of waste to be placed inside the can.
Specifications
Brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications) |