missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Belinostat

Description
Belinostat is a potent pan-HDAC inhibitor (IC50 = 28 nM). Dose-dependently reduces tumor volume in a rat model of glioblastoma and induces 70% and 28% apoptosis in LN-229 and LN-18 glioma cell lines (IC50 values are 210 nM and 300 nM respectively). In H358 cells, Belinostat dose-dependently reduces cell viability and upregulates amino acids and metabolites of the TCA and urea cycle. Also potently inhibits C4 release from stem cell-derived astrocytes and shows antidepressant activity. Belinostat is brain penetrant.
Specifications
Specifications
| Chemical Name or Material | (2E)-N-Hydroxy-3-[3-[(phenylamino)sulfonyl]phenyl]-2-propenamide |
| CAS | 866323-14-0 |
| Quantity | 10 mg |
| Target | Histone Deacetylase Inhibitors |
| Molecular Formula | C15H14N2O4S |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?