missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Belinostat
GREENER_CHOICE

Product Code. 30000421
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000421 50 mg 50mg
30000418 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30000421 Supplier Tocris Bioscience™ Supplier No. 7981/50
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent pan-HDAC inhibitor

Belinostat is a potent pan-HDAC inhibitor (IC50 = 28 nM). Dose-dependently reduces tumor volume in a rat model of glioblastoma and induces 70% and 28% apoptosis in LN-229 and LN-18 glioma cell lines (IC50 values are 210 nM and 300 nM respectively). In H358 cells, Belinostat dose-dependently reduces cell viability and upregulates amino acids and metabolites of the TCA and urea cycle. Also potently inhibits C4 release from stem cell-derived astrocytes and shows antidepressant activity. Belinostat is brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-N-Hydroxy-3-[3-[(phenylamino)sulfonyl]phenyl]-2-propenamide
CAS 866323-14-0
Quantity 50 mg
Target Histone Deacetylase Inhibitors
Molecular Formula C15H14N2O4S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.