missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Benchkote Surface Protector, 460 x 570mm pad

Product Code. 11979144
Change view
Click to view available options
Dimensions (L x W):
460 x 570 mm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11979144 460 x 570 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11979144 Supplier Cytiva Supplier No. 2300599
  • ONLINE EXCLUSIVE PRICE
    142.00 €
    130.33 € / Pack of 50
    Save 11.67 €
    8% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

An absorbent lab paper that protects surfaces against hazardous spills. After use, the bench protector sheet is incinerated or disposed of according to local regulations. Benchkote surface protector minimizes mess and contamination during spills.

  • Strong: Benchkote surface protector is strong and tear resistant when wet or dry
  • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
  • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Disposable: Benchkote paper can be incinerated after use
  • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
TRUSTED_SUSTAINABILITY

Specifications

Product Type Pad
Dimensions (L x W) 460 x 570 mm
Disposable Single-use
Quantity 50 Pads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.