missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Benchtop Arium pro VF TOC Ultrapure Water System
BIOPHARMA

Product Code. 14831392 Shop All Sartorius Products
Change view
Click to view available options
Conductivity:
100μS/cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conductivity Unit Size
14831392 100μS/cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 14831392 Supplier Sartorius Supplier No. H2OPROVFTTOC
  • ONLINE EXCLUSIVE PRICE
    12275.00 €
    9101.08 € / Pack of 1
    Save 3173.92 €
    25% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides widest selection of features and options for customised configuration.

  • Modular units pre-assembled according to specific applications
  • Lowest TOC level, along with real-time monitoring function
  • Efficient removal of endotoxins, DNase and RNase
  • High Flexibility
  • Total ease of operation
  • Meets and exceeds the ASTM Type 1 standard, ensuring reproducibility of results
  • Applications: HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC analysis, PCR, Electrophoresis, Endotoxin analysis, Immunocytochemistry, Pure water for preparation of cell culture media, Pure water for production of monoclonal antibodies, Photometry
TRUSTED_SUSTAINABILITY

Specifications

Conductivity 100μS/cm
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Distilled, DI, RO, EDI
Flow Rate 1.7 L/min.
Product Water Type Type 1
Remote Dispenser No
Resistivity 18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring Yes
Total Organic Carbon ≤2 ppb
DNase Yes
For Use With (Application) Cell Culture; Buffer and Media Prep; Histology; IC; Preparation of Reagents; Photometry; ELISA; AAS
Mounting Benchtop
Silica <1000ppb
Turbidity <1 NTU
Electrical Requirements 100/240 V
Voltage 100/240 V
Daily Output 100 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Arium pro Ultrapure Water System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.