Learn More
Jenway™ Benchtop Dissolved Oxygen Meter
Product Code. 12600493
Click to view available options
Voltage:
120 V
Unit Size:
Each
Product Code. 12600493
Brand: Jenway™ 3541702
Only null left
Add to basket
Add to basket
Store up to 250 DO data points and 20 BOD data sets.
  • LCD provides clear, direct readout of percentage of dissolved oxygen, mg/L, or BOD of samples
  • Easily export data to computer or printer via RS-232 or IrDA connections
  • % Saturation from 0 to 199 or 0 to 25 with accuracy of ±2% with 50°F (10°C) of calibration temperature
  • Manual salinity and barometric pressure compensation
  • Features real-time clock
Specifications
Data Points DO, temperature
Languages English, French, German, Italian, Spanish, Portuguese
Power Consumption 9 V power supply
Voltage 120 V