missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ Benchtop pH Meters
Description
- Supports one, two, or three point pH calibration
- Up to three decimal place resolution for accurate readings
Specifications
Specifications
| Description | Standard |
| Includes | Glass electrode, ATC, buffers |
| Accuracy (Temperature) | ±0.5°C/±1°F |
| Accuracy (pH) | ±0.003 |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Range (mV) | -1999.9 to 19.999 |
| Display Type | LCD, Custom with Backlight |
| Type | Standard Digital pH Meter Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?