missing translation for 'onlineSavingsMsg'
Learn More

Bendamustine hydrochloride, MedChemExpress

Product Code. 16478076
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
25mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478076 100 mg 100mg
16453786 25 mg 25mg
16441366 200 mg 200mg
16402006 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478076 Supplier Medchem Express Supplier No. HYB0077
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bendamustine hydrochloride (SDX-105), a purine analogue, is a DNA cross-linking agent. Bendamustine hydrochloride activats DNA-damage stress response and apoptosis. Bendamustine hydrochloride has potent alkylating, anticancer and antimetabolite properties.

Bendamustine hydrochloride (SDX-105), a purine analogue, is a DNA cross-linking agent. Bendamustine hydrochloride activats DNA-damage stress response and apoptosis. Bendamustine hydrochloride has potent alkylating, anticancer and antimetabolite properties.

Specifications

Chemical Name or Material Bendamustine hydrochloride
CAS 3543-75-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H22Cl3N3O2
Quantity 100 mg
Synonym SDX-105
Solubility Information DMSO : 100 mg/mL (253.34 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (31.67 mM; Need ultrasonic)
SMILES CN1C(C=CC(N(CCCl)CCCl)=C2)=C2N=C1CCCC(O)=O.Cl
Molecular Weight (g/mol) 394.72
Formula Weight 394.72
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.