missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bendazol, MedChemExpress
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Bendazol is a hypotensive drug which can also enhance NO synthase activity in renal glomeruli and collecting tubules.
Specifications
Specifications
| Chemical Name or Material | Bendazol |
| CAS | 621-72-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H12N2 |
| Quantity | 500 mg |
| Solubility Information | DMSO : 100 mg/mL (480.17 mM; Need ultrasonic) |
| SMILES | C1(CC2=CC=CC=C2)=NC3=CC=CC=C3N1 |
| Molecular Weight (g/mol) | 208.26 |
| Formula Weight | 208.26 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur