missing translation for 'onlineSavingsMsg'
Learn More

Benfotiamine, TRC

Product Code. 17973145
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973145 5 g 5g
17953245 100 mg 100mg
17921655 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973145 Supplier Toronto Research Chemicals Supplier No. TRCB1360005G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22457-89-2
Molecular Formula C19 H23 N4 O6 P S
Molecular Weight (g/mol) 466.45
Synonym Benzenecarbothioic acid, S-[2-[[(4-amino-2-methyl-5-pyrimidinyl)methyl]formylamino]-1-[2-(phosphonooxy)ethyl]-1-propen-1-yl] ester, Benzenecarbothioic acid, S-[2-[[(4-amino-2-methyl-5-pyrimidinyl)methyl]formylamino]-1-[2-(phosphonooxy)ethyl]-1-propenyl] ester (9CI), Benzoic acid, thio-, S-ester with N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-(4-hydroxy-2-mercapto-1-methyl-1-butenyl)formamide dihydrogen phosphate (ester) (8CI), Formamide, N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-(4-hydroxy-2-mercapto-1-methyl-1-butenyl)-, S-benzoate O-(dihydrogen phosphate) (8CI), 8088CB, BTMP, Benfothiamine, Benfotiamine, Benzoylthiamine O-monophosphate, Benzoylthiamine monophosphate, Berdi, Betivina, Bietamine, Biotamin, Milgamma, N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-(4-hydroxy-2-mercapto-1-methyl-1-butenyl)formamide S-benzoate O-phosphate, Neurostop, Nitanevril, S-Benzoylthiamine O-monophosphate, S-Benzoylthiamine monophosphate, Tabiomyl, Vitanevril, (E)-6-(4-Amino-2-methylpyrimidin-5-yl)-3-benzoylthio-5-formyl-4-methyl-5-azahex-3-enyldihydrogenphosphat (ABDA Nomenklatur)
IUPAC Name S-[(E)-2-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-5-phosphonooxypent-2-en-3-yl] benzenecarbothioate
SMILES C\C(=C(\CCOP(=O)(O)O)/SC(=O)c1ccccc1)\N(Cc2cnc(C)nc2N)C=O

Specifications

InChI Formula InChI=1S/C19H23N4O6PS/c1-13(23(12-24)11-16-10-21-14(2)22-18(16)20)17(8-9-29-30(26,27)28)31-19(25)15-6-4-3-5-7-15/h3-7,10,12H,8-9,11H2,1-2H3,(H2,20,21,22)(H2,26,27,28)/b17-13+
Formula Weight 466.1076
Percent Purity >95%
Quantity 5 g
Chemical Name or Material Benfotiamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.