missing translation for 'onlineSavingsMsg'
Learn More

Benidipine hydrochloride, MedChemExpress

Product Code. 16428766
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428766 Supplier Medchem Express Supplier No. HYB1448
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benidipine hydrochloride is a dihydropyridine calcium channel blocker for the treatment of high blood pressure (hypertension).

Benidipine hydrochloride is a dihydropyridine calcium channel blocker for the treatment of high blood pressure (hypertension).

Specifications

Chemical Name or Material Benidipine hydrochloride
CAS 91599-74-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H32ClN3O6
Quantity 100 mg
Synonym KW-3049
Solubility Information DMSO : 33.33 mg/mL (61.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(C)NC(C)=C(C(O[C@H]2CN(CC3=CC=CC=C3)CCC2)=O)[C@@H]1C4=CC=CC([N+]([O-])=O)=C4)OC.[H]Cl
Molecular Weight (g/mol) 542.02
Formula Weight 542.02
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.