missing translation for 'onlineSavingsMsg'
Learn More

Benorylate, Thermo Scientific Chemicals

Product Code. 16697052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16697052 5 g 5g
16607062 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16697052 Supplier Thermo Scientific Acros Supplier No. 465960050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5003-48-5
Molecular Formula C17H15NO5
Molecular Weight (g/mol) 313.31
MDL Number MFCD00864257
InChI Key FEJKLNWAOXSSNR-UHFFFAOYSA-N
Synonym Benorilate
IUPAC Name 4-acetamidophenyl 2-(acetyloxy)benzoate
SMILES CC(=O)NC1=CC=C(OC(=O)C2=CC=CC=C2OC(C)=O)C=C1

Specifications

Alpha Vector BENORYLATE 001
Melting Point 177°C to 182°C
Color White
Loss on Drying ≤0.5 %
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
Formula Weight 313.31
Physical Form Crystalline Powder
Chemical Name or Material Benorylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.