Learn More
Lenz Laborglasinstrumente™ Adapter with NS Stopcock, Bent
Product Code. 10359860
Change view
Click to view available options
Standard Taper Joint:
NS 19/26
NS 24/29
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10359860 NS 24/29 Each
10460513 NS 19/26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10359860 Supplier Lenz Laborglasinstrumente™ Supplier No. 05098003
  • ONLINE EXCLUSIVE PRICE
    48.36 €
    37.08 € / Pack of 1
    Save 11.28 €
    23% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
For distillation tubing connections. Lenz Laborglasinstrumente™ Adapter with NS Stopcock, Bent are manufactured from durable DURAN™ glass tubing.
  • Bent glass olive at 90° angle
  • Bore: 2.5mm
  • Stopcock with solid glass plug
  • Temperature and chemical resistance
Specifications
Type Bent Adapter with NS Stopcock
Material Duran Glass
Standard Taper Joint NS 24/29