Learn More
Lenz Laborglasinstrumente™ Bent Receiver Adapter with Glass Olive
missing translation for 'orderingAttributeHoverText'
Standard Taper Joint:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
missing translation for 'unitSize'
Each
Description
- NS socket and NS cone
- 105° angle
- Vacuum connection
- Glass olive with drain tip
- Meets DIN 12594 standards
Specifications
Specifications
| Type | Bent Receiver Adapter with Glass Olive |
| Material | Duran Glass |
| Standard Taper Joint | NS 19/26 |