missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bentazon-d7, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Bentazon-d7 |
| CAS | 131842-77-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 D7 H5 N2 O3 S |
| Quantity | 1 mg |
| Synonym | Bentazone D7 (isopropyl D7), 1H-2,1,3-Benzothiadiazin-4(3H)-one, 3-[1-(methyl-d3)ethyl-1,2,2,2-d4]-, 2,2-dioxide (9CI), 3-Isopropyl-d7-1H-2,1,3-benzothiadiazin-4(3H)-one 2,2-dioxide |
| InChI Formula | InChI=1S/C10H12N2O3S/c1-7(2)12-10(13)8-5-3-4-6-9(8)11-16(12,14)15/h3-7,11H,1-2H3/i1D3,2D3,7D |
| SMILES | [2H]C([2H])([2H])C([2H])(N1C(=O)c2ccccc2NS1(=O)=O)C([2H])([2H])[2H] |
| IUPAC Name | 3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-2,2-dioxo-1H-2λ^{6},1,3-benzothiadiazin-4-one |
| Molecular Weight (g/mol) | 247.322 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction