missing translation for 'onlineSavingsMsg'
Learn More

Bentiromide Sodium Salt, TRC

Product Code. 30340141
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340141 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340141 Supplier Toronto Research Chemicals Supplier No. TRCB13046550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bentiromide Sodium Salt
CAS 41748-47-4
Molecular Formula C23H19N2NaO5
Quantity 50 mg
Synonym Benzoic acid, 4-[[2-(benzoylamino)-3-(4-hydroxyphenyl)-1-oxopropyl]amino]-, monosodium salt, (S)- (9CI), N-Benzoyl-L-tyrosyl-p-aminobenzoic acid sodium salt, Sodium N-benzoyl-L-tyrosyl-p-aminobenzoate, Bentiromide sodium, Bentiromide sodium salt, 4-[[(2S)-2-(Benzoylamino)-3-(4-hydroxyphenyl)-1-oxopropyl]amino]benzoic acid monosodium salt
InChI Formula InChI=1S/C23H20N2O5.Na/c26-19-12-6-15(7-13-19)14-20(25-21(27)16-4-2-1-3-5-16)22(28)24-18-10-8-17(9-11-18)23(29)30;/h1-13,20,26H,14H2,(H,24,28)(H,25,27)(H,29,30);/q;+1/p-1/t20-;/m0./s1
SMILES O=C(N[C@H](C(NC1=CC=C(C(O[Na])=O)C=C1)=O)CC2=CC=C(O)C=C2)C3=CC=CC=C3
IUPAC Name sodium;4-[[(2S)-2-benzamido-3-(4-hydroxyphenyl)propanoyl]amino]benzoate
Molecular Weight (g/mol) 426.4
Formula Weight 426.1192
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.