missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, Honeywell Fluka™

Product Code. 15672930 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15672930 500 g Poly Bottle 500g
15662930 2.5 kg Drum with inliner 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15672930 Supplier Honeywell-Fluka Supplier No. 285234500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

pH 7.0-10.5 (2 %, 20°C)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-78-9
MDL Number MFCD00130611

Specifications

Packaging Poly Bottle
Quantity 500 g
UN Number NONH for all modes of transport
Chemical Name or Material Bentonite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.